missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pelican™ 1200 Small Case
Lightweight cas is extremely strong, compact, and fully capable.
Supplier: Pelican™ 1200 000 110
Description
- Watertight, crushproof, and dustproof
- Easy open Double Throw latches
- Open cell core with solid wall design makes the case strong yet light weight
- O-ring seal
- Automatic Pressure Equalization Valve balances interior pressure, keeps water out
- Stainless steel hardware
- Pick N Pluck™ with convoluted lid foam
- Lifetime Guarantee of Excellence
Certifications
IP67/TANAG 4280/Def Stan 81-41
Chemical Identifiers
| Watertight Equipment Case | |
| 4.87 in. | |
| 12.4 cm | |
| 10.62 x 9.68 in. (27 x 24.6 cm) | |
| 10.62 in. | |
| 27 cm | |
| 1200 | |
| 1.2 kg | |
| 7.12 in. | |
| 18.1 cm |
| Polypropylene, Polyurethane Foam | |
| 4.12 in. | |
| 10.5 cm | |
| Pick N' Pluck | |
| 9.25 in. | |
| 23.5 cm | |
| 2.45 lb. | |
| 9.68 in. | |
| 24.6 cm |
Specifications
| Watertight Equipment Case | |
| Polypropylene, Polyurethane Foam | |
| 4.12 in. | |
| 10.5 cm | |
| Pick N' Pluck | |
| 9.25 in. | |
| 23.5 cm | |
| 2.45 lb. | |
| 9.68 in. | |
| 24.6 cm |
| Black | |
| 4.87 in. | |
| 12.4 cm | |
| 10.62 x 9.68 in. (27 x 24.6 cm) | |
| 10.62 in. | |
| 27 cm | |
| 1200 | |
| 1.2 kg | |
| 7.12 in. | |
| 18.1 cm |