Learn More
New England Biolab Ltd SHuffle T7 Express lysY Competent E.coli 12 x 0.05 ml/tube DFS Item
Supplier:  New England Biolab Ltd C3030J
Chemically competent E. coli cells suitable for T7 protein expression with enhanced capacity to correctly fold proteins with multiple disulfide bonds in the cytoplasm and with enhanced reduction of basal expression.
Catalog No. NC1270293
Only null left
Explore available promotions