Learn More
Ibidi Gmbh 12 Well Chamber, removable
Catalog No. 50305813
Encompass
The 12 Well Chamber, removable is a removable silicone chamber, mounted on a glass slide, for cell culture and immunofluorescence staining. It allows the use of standard cultivation, staining and mounting techniques with coverslip sealing. After mounting the glass slide with a coverslip, it is ready for the use with upright and inverted microscopes, as well as for long term storage. Suitable 24 mm60 mm coverslips are provided by ibidi (Cat.# 10811). Glass slide, silicone chamber, sterile, 15 pcs./box
Catalog No. 50305813 Supplier Ibidi Gmbh Supplier No. 81201
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The 12 Well Chamber, removable is a removable silicone chamber, mounted on a glass slide, for cell culture and immunofluorescence staining. It allows the use of standard cultivation, staining and mounting techniques with coverslip sealing. After mounting the glass slide with a coverslip, it is ready for the use with upright and inverted microscopes, as well as for long term storage. Suitable 24 mm60 mm coverslips are provided by ibidi (Cat.# 10811). Glass slide, silicone chamber, sterile, 15 pcs./box