missing translation for 'onlineSavingsMsg'
Learn More

12-Molybdophosphoric acid hydrate, Mo >47.5%

Catalog No. p-7046697
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1582714 25 g
AAL1582722 100 g
AAL1582736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1582714 Supplier Thermo Scientific Chemicals Supplier No. L1582714
Only null left

CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol

12-Molybdophosphoric acid hydrate is used as a precursor to hybrid organic-inorganic based molecular batteries with application as cation-insertion electrode. It finds application as an analytical reagent in thin layer chromatography for staining phenolics, hydrocarbon waxes, alkaloids and steroids. It acts as a reactant for the preparation of catalysts for the nitration of aromatic compounds, highly dispersed MoOx/CNTs (carbon nanotubes) as a support for Pt catalysts and anchored Wilkinson catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
12-Molybdophosphoric acid hydrate is used as a precursor to hybrid organic-inorganic based molecular batteries with application as cation-insertion electrode. It finds application as an analytical reagent in thin layer chromatography for staining phenolics, hydrocarbon waxes, alkaloids and steroids. It acts as a reactant for the preparation of catalysts for the nitration of aromatic compounds, highly dispersed MoOx/CNTs (carbon nanotubes) as a support for Pt catalysts and anchored Wilkinson catalyst.

Solubility
Soluble in water, ethanol and ether.

Notes
Hygroscopic. Incompatible with organic materials and powdered metals.

Chemical Identifiers

CAS 51429-74-4
Molecular Formula H3Mo12O40P
Molecular Weight (g/mol) 1825.40
MDL Number MFCD00149913
InChI Key FEFSFHGZSNHJEL-UHFFFAOYSA-N
Synonym Molybdophosphoric acid
IUPAC Name phosphoric acid dodecamolybdenum hexatriacontaoxidandiide
SMILES [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].OP(O)(O)=O

Specifications

Melting Point 78°C to 90°C
Density 3.15
Sensitivity Hygroscopic
Quantity 25 g
UN Number UN3085
Merck Index 14,7342
Solubility Information Soluble in water,ethanol and ether.
Formula Weight 1825.25 (Anhydrous)
Assay Mo >47.5%
Chemical Name or Material 12-Molybdophosphoric acid hydrate
Hazard Category H272-H290-H314-H335-H373
Hazard Statement GHS H Statement
H272-H314-H318
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P220-P221-P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 234-713-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only