missing translation for 'onlineSavingsMsg'
Learn More

Brady™ 12-Lock Lockout Station Polypropylene Unfilled

Catalog No. 17607631
Change view
Click to view available options
Depth (English):
3.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English)
17607631 3.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17607631 Supplier Brady™ Supplier No. 170397

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Safely power down faster when you power up your lockout tagout readiness with this easily installed, accessible lockout station.

Safely power down faster when you power up your lockout tagout readiness with this easily installed, accessible lockout station.

  • OSHA compliance driver, designed exclusively for safety to help you comply with OSHA 29 CFR 1910.147 Control of Hazardous Energy.
  • Built to last, all components meet Brady's highest standards for quality, durability and reliability.
  • Ultimate accessibility, place locks, tags and devices at the point of need to help authorized workers maintain safety and compliance.
  • Comes with predrilled holes for easy wall mounting when you want it to stay put and carrying handle when you need it on the go.
  • 16 in. H x 14 in. W x 3.5 in. D station holds up to 12 padlocks plus lockout hasps and tags.
  • Lockable clear plastic door provides instant visibility while protecting contents from damage, theft and loss.
  • Lockout devices sold separately.

Specifications

Product Type Lockout Station Unfilled
Material Polypropylene
Depth (English) 3.5 in.
Depth (Metric) 88.90 mm
Description Unfilled
Height (English) 16 in.
Height (Metric) 406.40 mm
Includes (1) Carrying Handle, (6) Lock Hooks (can hold 2 padlocks), (2) Tag Holders, (1) Device Bin, Trillingual Labels
Width (English) 14 in.
Width (Metric) 355.60 mm