missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ High Voltage Leather Protectors

Catalog No. 17001633
Change view
Click to view available options
Size:
10
11
12
8
9
Length (English):
10 in.
12 in.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English)
17001633 8 10 in.
17001668 8 12 in.
17001634 9 10 in.
17001669 9 12 in.
17001635 10 10 in.
17001670 10 12 in.
17001636 11 10 in.
17001671 11 12 in.
17001637 12 10 in.
17001672 12 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17001633 Supplier National Safety Apparel™ Supplier No. DWH10L8

Glove protectors should be used with electrical insulating rubber gloves for additional protection against mechanical hazards.

  • Leather glove protectors should be used every time that a task requires electrical insulating gloves
  • 10 in. glove protectors are suitable for use with Class 0 to 00 electrical insulating rubber gloves
  • 12 in. glove protectors are suitable for use with Class 2 electrical insulating rubber gloves

Specifications

Certifications/Compliance ASTM F696, NFPA 70E, CSA Z462, OSHA 1910.269
Applications Industrial
Color White, Yellow
Hand Specific
Material Leather
Product Type Glove Protectors
Size 8
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
For Use With (Equipment) Class 00 to 0 Electrical Insulating Rubber Gloves
Glove Cuff Style Elasticated
Series DWH10
Length (English) 10 in.
Length (Metric) 25.4 cm
Voltage 1000 V
Show More Show Less