missing translation for 'onlineSavingsMsg'
Learn More

[12]Cycloparaphenylene 90.0+%, TCI America™

Catalog No. C244910MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C244910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C244910MG Supplier TCI America Supplier No. C244910MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1092522-75-2
Molecular Formula C72H48
Molecular Weight (g/mol) 913.176
InChI Key AAFTYBVDGIFJMP-UHFFFAOYSA-N
Synonym [12]CPP
PubChem CID 53384443
SMILES C1=CC2=CC=C1C3=CC=C(C=C3)C4=CC=C(C=C4)C5=CC=C(C=C5)C6=CC=C(C=C6)C7=CC=C(C=C7)C8=CC=C(C=C8)C9=CC=C(C=C9)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C2C=C1

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 913.18
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [12]Cycloparaphenylene