missing translation for 'onlineSavingsMsg'
Learn More

12-Crown 4-Ether 95.0+%, TCI America™

Catalog No. p-7124648
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08581ML 1 mL
C08585ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08581ML Supplier TCI America Supplier No. C08581ML
Only null left

Chemical Identifiers

CAS 294-93-9
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00005103
InChI Key XQQZRZQVBFHBHL-UHFFFAOYSA-N
PubChem CID 9269
ChEBI CHEBI:32399
IUPAC Name 1,4,7,10-tetraoxacyclododecane
SMILES C1COCCOCCOCCO1

Specifications

Color Colorless
Boiling Point 70°C
Quantity 1 mL
UN Number 2810
Synonym 12-crown-4, eoct, 12-crown-4 ether, ethylene oxide cyclic tetramer, ccris 157, unii-k6069p2c2a, 12-crown4-ether, 12-crown 4-ether, 5-19-11-00334 beilstein handbook reference, 3,6,9,12-tetraoxacyclododecan
Formula Weight 176.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 12-Crown 4-Ether
RTECSNumber XF0550000
TSCA Yes