missing translation for 'onlineSavingsMsg'
Learn More

12-Crown-4, 98%

Catalog No. aaa1197206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1197206 5 g
AAA1197203 1 g
AAA1197214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1197206 Supplier Thermo Scientific Chemicals Supplier No. A1197206
Only null left

CAS: 294-93-9 | C8H16O4 | 176.21 g/mol

It serves as an aprotic solvent for alkali metals, the sodium solution is a strong reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It serves as an aprotic ″solvent″ for alkali metals, the sodium solution is a strong reducing agent.

Solubility
Fully miscible in water.

Notes
Light & Air Sensitive & Hygroscopic. Store under inert gas. Keep in a tightly closed container.

Chemical Identifiers

CAS 294-93-9
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00005103
InChI Key XQQZRZQVBFHBHL-UHFFFAOYSA-N
PubChem CID 9269
ChEBI CHEBI:32399
IUPAC Name 1,4,7,10-tetraoxacyclododecane
SMILES C1COCCOCCOCCO1

Specifications

Melting Point 17°C
Density 1.108
Boiling Point 68°C to 70°C (0.1 mmHg)
Flash Point >109°C (228°F)
Odor Odorless
Refractive Index 1.462
Quantity 5 g
UN Number UN2810
Beilstein 1363064
Sensitivity Air and light sensitive; Hygroscopic
Synonym 12-crown-4, eoct, 12-crown-4 ether, ethylene oxide cyclic tetramer, ccris 157, unii-k6069p2c2a, 12-crown4-ether, 12-crown 4-ether, 5-19-11-00334 beilstein handbook reference, 3,6,9,12-tetraoxacyclododecan
Solubility Information Fully miscible in water.
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material 12-Crown-4
Show More Show Less
Hazard Category H330
Hazard Statement GHS H Statement
H330-H361-H315-H319
Fatal if inhaled.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P271-P284-P304+P340-P310-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 206-036-5
RTECSNumber XF0550000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only