missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 12 cal Coverall

Catalog No. p-7220106
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17001541 Small
17001542 Medium
17001543 Large
17001544 X-Large
17001545 2X-Large
17001546 3X-Large
17001547 4X-Large
17001548 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17001541 Supplier National Safety Apparel™ Supplier No. C88UPSM32
Only null left

Durable and comfortable ultrasoft coveralls offer a unique blend of 88% cotton and 12% high-tenacity nylon for a smooth feel. The inherently FR material delivers exceptional performance and reliability.

  • Comfortable sewn-in elastic waist for better fit
  • Dual certified to protect against flash fire and arc flash hazards
  • 2-way covered zipper front for easy entry
  • Two front chest pockets with flaps and hidden snap closures
  • Large reinforced back pockets
  • Convenient 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • All metal covered to ensure no contact with skin
  • Action back allows for full range of motion
  • FR fabric will not melt, drip, or ignite
  • Zip front closure with additional covered snaps

Specifications

Product Type Coveralls
Size Small
Gender Unisex
Material Cotton, Nylon
Color Navy
Disposable Reusable
Waist Style Elastic (Stitched-in)
Ankle Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Protection Class CAT 2
Resistance Flame
Arc Flash Rating 12 cal/cm²
Certifications/Compliance ASTM F1506, OSHA 1910.269, NFPA 70E, NFPA 2112, CSA Z462
Suspender None
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Style Coveralls
Show More Show Less