missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ 12 Cal UltraSoft FR Coveralls 28 in. Inseam

Catalog No. p-7220107
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17001525 Small
17001526 Medium
17001527 Large
17001528 X-Large
17001529 2X-Large
17001530 3X-Large
17001531 4X-Large
17001532 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17001525 Supplier National Safety Apparel™ Supplier No. C88UPSM28
Only null left

For enhanced protection Westex™ UltraSoft is the premium choice for superior comfort, improved safety and above average durability. With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability

  • Arc Rating: 12 cal/cm²
  • CAT 2
  • Fabric/Material: 9 oz. Westex™ UltraSoft™
  • Comfortable sewn-in elastic waist for better fit
  • Dual certified to protect against flash fire and arc flash hazards
  • 2-way covered zipper front for easy entry
  • Two front chest pockets with flaps and hidden snap closures
  • Large reinforced back pockets
  • Convenient 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • All metal covered to ensure no contact with skin
  • Action back allows for full range of motion
  • FR fabric will not melt, drip, or ignite
  • Zip front closure with additional covered snaps
  • Home laundry
  • Made in the USA

Specifications

Size Small
Material Cotton, Nylon
Color Navy
Ankle Style Open
Closure Type Zipper
Seam Type Stitched
Certifications/Compliance Arc Rating: 12 cal/cm², CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, NFPA 2112, CSA Z462
Suspender None