missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ 16 cal Arc Goggle Hood System

Catalog No. 170012006
Encompass
Change view
Click to view available options
Arc Flash Rating:
12 cal/cm²
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash Rating
170012006 12 cal/cm²
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 170012006 Supplier National Safety Apparel™ Supplier No. KITHP12GGL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kit includes: Navy UltraSoft™ Balaclava, Standard Arc Goggle, Elastic Strap to Secure Goggles

  • Lightweight alternative for 16 cal with 360 degree head protection
  • Provides increased flexibility with excellent breathability
  • Goggles feature baffled top and bottom vents for increased airflow
  • Anti-fog inner lens
  • Full facial protection extends below neckline
  • Stretch material keeps head and neck comfortable
  • Nose covering
  • Made in the USA

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 2, NFPA 70E, ASTM F2178, OSHA 1910.269, ANSI/ISEA Z87.1
Closure Type None
Product Type Goggles
Size Universal
Material Westex™ UltraSoft™, OPF Para-Aramid/OPF Blend
Arc Flash Rating 12 cal/cm²
Suspender None
Color Navy
Seam Type Stitched
Show More Show Less