missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ 12 Cal ArcGuard™ UltraSoft Arc Flash Leggings

Catalog No. p-7220296
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170012047 Large
170012048 X-Large
170012049 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 170012047 Supplier National Safety Apparel™ Supplier No. L05UPLG
Only null left

Ultrasoft flame resistant leggings are comfortable and typically paired with an arc rated long coat. Compliant to NFPA 70E with an arc rating of 12 cal/cm² and an CAT level 2.

  • 9 oz. UltraSoft™
  • Adjustable FR hook and loop closure
  • Single layer legging with foot flap
  • For use with 12 cal/cm2 FR long coat

Specifications

Size Large
Gender Unisex
Material Cotton, Nylon
Color Navy
Ankle Style Open
Seam Type Stitched
Disposable Reusable
Arc Flash Rating 12 cal/cm²
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, CSA Z462
Sterility Non-sterile