missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ 12 Cal ArcGuard™ UltraSoft Arc Flash Leggings

Catalog No. 170012047
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170012047 Large
170012048 X-Large
170012049 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 170012047 Supplier National Safety Apparel™ Supplier No. L05UPLG

Ultrasoft flame resistant leggings are comfortable and typically paired with an arc rated long coat. Compliant to NFPA 70E with an arc rating of 12 cal/cm² and an CAT level 2.

  • 9 oz. UltraSoft™
  • Adjustable FR hook and loop closure
  • Single layer legging with foot flap
  • For use with 12 cal/cm2 FR long coat

Specifications

Size Large
Gender Unisex
Material Cotton, Nylon
Color Navy
Ankle Style Open
Seam Type Stitched
Disposable Reusable
Arc Flash Rating 12 cal/cm²
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, CSA Z462
Sterility Non-sterile