missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ 12 Cal ArcGuard™ Arc Flash Kit with Short Coat, Bib Overall with Lift Front Hood (08)

Catalog No. 170011289
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011289 5X-Large
170011254 Small
170011259 Medium
170011264 Large
170011269 X-Large
170011274 2X-Large
170011279 3X-Large
170011284 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 170011289 Supplier National Safety Apparel™ Supplier No. KIT2SC11LF5X08

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability

  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • FR hook and loop front closure on short coat
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Hook and loop at side of bib for easy access
  • Expandable leg opening to fit over boots
  • Made in the USA

Specifications

Certifications/Compliance CAT 2, NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Material Cotton, Nylon
Arc Flash Rating 12 cal/cm²
Size 5X-Large
Closure Type FR Hook and Loop
Color Navy
Ankle Style Open
Suspender None
Seam Type Stitched
Show More Show Less