missing translation for 'onlineSavingsMsg'
Learn More

12-Bromo-1-dodecanol 98.0+%, TCI America™

Catalog No. p-7119230
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17315G 5 g
B173125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17315G Supplier TCI America Supplier No. B17315G

Chemical Identifiers

CAS 3344-77-2
Molecular Formula C12H25BrO
Molecular Weight (g/mol) 265.24
MDL Number MFCD00004754
InChI Key ASIDMJNTHJYVQJ-UHFFFAOYSA-N
PubChem CID 137895
IUPAC Name 12-bromododecan-1-ol
SMILES OCCCCCCCCCCCCBr

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 156°C
Quantity 5 g
Formula Weight 265.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 12-Bromo-1-dodecanol
TSCA No
Recommended Storage Refrigerator