Learn More
12-Azidododecylphosphonic acid, 95%
CAS: 721457-32-5 | C12H26N3O3P | 291.332 g/mol
$278.92 - $755.55
Chemical Identifiers
| CAS | 721457-32-5 |
|---|---|
| Molecular Formula | C12H26N3O3P |
| Molecular Weight (g/mol) | 291.332 |
| MDL Number | MFCD17012690 |
| InChI Key | WHRLXOVKIALVOF-UHFFFAOYSA-N |
| PubChem CID | 97638952 |
| IUPAC Name | 12-azidododecylphosphonic acid |
| SMILES | C(CCCCCCP(=O)(O)O)CCCCCN=[N+]=[N-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH33784MD
|
Thermo Scientific Chemicals
H33784MD |
250 mg |
Each for $278.92
|
|
|||||
|
AAH3378403
|
Thermo Scientific Chemicals
H3378403 |
1 g |
Each for $755.55
|
|
|||||
Description
This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThis coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.
Notes
Store in refrigerator. Protect from heat. Incompatible with oxidizing agents.
Chemical Identifiers
| 721457-32-5 | |
| 291.332 | |
| WHRLXOVKIALVOF-UHFFFAOYSA-N | |
| 12-azidododecylphosphonic acid |
| C12H26N3O3P | |
| MFCD17012690 | |
| 97638952 | |
| C(CCCCCCP(=O)(O)O)CCCCCN=[N+]=[N-] |
Specifications
| 721457-32-5 | |
| C12H26N3O3P | |
| MFCD17012690 | |
| WHRLXOVKIALVOF-UHFFFAOYSA-N | |
| 12-azidododecylphosphonic acid | |
| 97638952 | |
| 95% |
| 83°C to 85°C | |
| N3(CH2)12P(O)(OH)2 | |
| 250 mg | |
| C(CCCCCCP(=O)(O)O)CCCCCN=[N+]=[N-] | |
| 291.332 | |
| 291.33 | |
| 12-Azidododecylphosphonic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only