missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11alpha-Hydroxyprogesterone 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 169°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 330.47 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 11alpha-Hydroxyprogesterone |
Safety and Handling
| RTECSNumber | TU5057000 |
| TSCA | No |