Learn More
Mopec™ #11 Sterile Scalpel Blades
Description
- Individually wrapped
- Fits #3, #3L and #5 scalpel handles
Specifications
Specifications
| Blade Material | Stainless Steel |
| Product Type | Scalpel |
| Size | # 11 |
| Disposable | Single-use |
| Sterility | Sterile |
| Quantity | 100 Blades/Pk. |
| Graduated | No |