Learn More
11-Hydroxyundecylphosphonic acid, 95%
CAS: 83905-98-0 | C11H25O4P | 252.291 g/mol
$300.10 - $824.40
Chemical Identifiers
| CAS | 83905-98-0 |
|---|---|
| Molecular Formula | C11H25O4P |
| Molecular Weight (g/mol) | 252.291 |
| MDL Number | MFCD11982869 |
| InChI Key | PPCDEFQVKBXBPS-UHFFFAOYSA-N |
| Synonym | 11-hydroxyundecyl phosphonic acid, 11-hydroxyundecyl-1-phosphonic acid, 11-hydroxyundecylphosphonic acid, phosphonic acid, 11-hydroxyundecyl, 11-hydroxyundecylphosphonic acid gc |
| PubChem CID | 45158971 |
| IUPAC Name | 11-hydroxyundecylphosphonic acid |
| SMILES | C(CCCCCO)CCCCCP(=O)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH33647MD
|
Thermo Scientific Chemicals
H33647MD |
250 mg |
Each for $300.10
|
|
|||||
|
AAH3364703
|
Thermo Scientific Chemicals
H3364703 |
1 g |
Each for $824.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 83905-98-0 | |
| 252.291 | |
| PPCDEFQVKBXBPS-UHFFFAOYSA-N | |
| 45158971 | |
| C(CCCCCO)CCCCCP(=O)(O)O |
| C11H25O4P | |
| MFCD11982869 | |
| 11-hydroxyundecyl phosphonic acid, 11-hydroxyundecyl-1-phosphonic acid, 11-hydroxyundecylphosphonic acid, phosphonic acid, 11-hydroxyundecyl, 11-hydroxyundecylphosphonic acid gc | |
| 11-hydroxyundecylphosphonic acid |
Specifications
| 83905-98-0 | |
| C11H25O4P | |
| MFCD11982869 | |
| 11-hydroxyundecyl phosphonic acid, 11-hydroxyundecyl-1-phosphonic acid, 11-hydroxyundecylphosphonic acid, phosphonic acid, 11-hydroxyundecyl, 11-hydroxyundecylphosphonic acid gc | |
| C(CCCCCO)CCCCCP(=O)(O)O | |
| 252.291 | |
| 252.29 | |
| 11-Hydroxyundecylphosphonic acid |
| 110°C to 112°C | |
| HO(CH2)11P(O)(OH)2 | |
| 250 mg | |
| PPCDEFQVKBXBPS-UHFFFAOYSA-N | |
| 11-hydroxyundecylphosphonic acid | |
| 45158971 | |
| 95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only