missing translation for 'onlineSavingsMsg'
Learn More

11-Hydroxyundecylphosphonic acid, 95%

Catalog No. AAH33647MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33647MD 250 mg
AAH3364703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33647MD Supplier Thermo Scientific Chemicals Supplier No. H33647MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 83905-98-0 | C11H25O4P | 252.291 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83905-98-0
Molecular Formula C11H25O4P
Molecular Weight (g/mol) 252.291
MDL Number MFCD11982869
InChI Key PPCDEFQVKBXBPS-UHFFFAOYSA-N
Synonym 11-hydroxyundecyl phosphonic acid, 11-hydroxyundecyl-1-phosphonic acid, 11-hydroxyundecylphosphonic acid, phosphonic acid, 11-hydroxyundecyl, 11-hydroxyundecylphosphonic acid gc
PubChem CID 45158971
IUPAC Name 11-hydroxyundecylphosphonic acid
SMILES C(CCCCCO)CCCCCP(=O)(O)O

Specifications

Melting Point 110°C to 112°C
Linear Formula HO(CH2)11P(O)(OH)2
Quantity 250 mg
Formula Weight 252.29
Percent Purity 95%
Chemical Name or Material 11-Hydroxyundecylphosphonic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only