missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11-Heneicosanone 95.0+%, TCI America™
$444.72 - $1168.57
Chemical Identifiers
| CAS | 19781-72-7 |
|---|---|
| Molecular Formula | C21H42O |
| Molecular Weight (g/mol) | 310.566 |
| MDL Number | MFCD00048410 |
| InChI Key | DOACSXJVHDTDSG-UHFFFAOYSA-N |
| Synonym | Didecyl Ketone |
| PubChem CID | 29776 |
| IUPAC Name | henicosan-11-one |
| SMILES | CCCCCCCCCCC(=O)CCCCCCCCCC |
Chemical Identifiers
| 19781-72-7 | |
| 310.566 | |
| DOACSXJVHDTDSG-UHFFFAOYSA-N | |
| 29776 | |
| CCCCCCCCCCC(=O)CCCCCCCCCC |
| C21H42O | |
| MFCD00048410 | |
| Didecyl Ketone | |
| henicosan-11-one |
Specifications
| 19781-72-7 | |
| White-Yellow | |
| MFCD00048410 | |
| Didecyl Ketone | |
| CCCCCCCCCCC(=O)CCCCCCCCCC | |
| 310.566 | |
| 310.57 | |
| Crystalline Powder |
| 64°C | |
| C21H42O | |
| 5 g | |
| DOACSXJVHDTDSG-UHFFFAOYSA-N | |
| henicosan-11-one | |
| 29776 | |
| ≥95.0% (GC) | |
| 11-Heneicosanone |
Safety and Handling
TSCA : No