missing translation for 'onlineSavingsMsg'
Learn More

11-Heneicosanol, 97%

Catalog No. AAL0518703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0518703 1 g
AAL0518706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0518703 Supplier Thermo Scientific Chemicals Supplier No. L0518703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3381-26-8 | C21H44O | 312.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3381-26-8
Molecular Formula C21H44O
Molecular Weight (g/mol) 312.582
MDL Number MFCD00026547
InChI Key BCNCKKYOXRHQGT-UHFFFAOYSA-N
Synonym 11-heneicosanol, 11-henicosanol, 11-henicosanol #, acmc-20aozg
PubChem CID 76913
IUPAC Name henicosan-11-ol
SMILES CCCCCCCCCCC(CCCCCCCCCC)O

Specifications

Melting Point 71°C to 73°C
Odor Odorless
Quantity 1 g
Beilstein 1765841
Formula Weight 312.58
Percent Purity 97%
Chemical Name or Material 11-Heneicosanol
EINECSNumber 222-184-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only