missing translation for 'onlineSavingsMsg'
Learn More

11-Bromoundecanol, 97%

Catalog No. p-7058418
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1444806 5 g
AAL1444814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1444806 Supplier Thermo Scientific Chemicals Supplier No. L1444806
Only null left

CAS: 1611-56-9 | C11H23BrO | 251.21 g/mol

11-Bromoundecanol was used in the preparation of 11-mercapto-1-undecanol1 and 1-(2-acryloyloxyundecyl)-3-methylimidazolium bromide. It was also used in one-pot synthesis of graft copolymers based on a metathesis skeleton with poly(methyl methacrylate) grafts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
11-Bromoundecanol was used in the preparation of 11-mercapto-1-undecanol1 and 1-(2-acryloyloxyundecyl)-3-methylimidazolium bromide. It was also used in one-pot synthesis of graft copolymers based on a metathesis skeleton with poly(methyl methacrylate) grafts.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1611-56-9
Molecular Formula C11H23BrO
Molecular Weight (g/mol) 251.21
MDL Number MFCD00004752
InChI Key XFGANBYCJWQYBI-UHFFFAOYSA-N
Synonym 11-bromo-1-undecanol, 11-bromoundecanol, 1-undecanol, 11-bromo, 1-bromo-11-hydroxyundecane, undecamethylene bromohydrin, 11-bromoundecyl alcohol, 11-bromo-l-undecanol, 11-bromoundecane-1-ol, 11-bromanylundecan-1-ol, acmc-1br3a
PubChem CID 74163
IUPAC Name 11-bromoundecan-1-ol
SMILES OCCCCCCCCCCCBr

Specifications

Melting Point 46°C to 49°C
Boiling Point 165°C to 170°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1741180
Solubility Information Insoluble in water.
Formula Weight 251.21
Percent Purity 97%
Chemical Name or Material 11-Bromoundecanol
Hazard Category H315-H319-H332-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-554-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only