missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11-Bromoundecanoic Acid 97.0+%, TCI America™
$84.70 - $947.18
Chemical Identifiers
| CAS | 2834-05-1 |
|---|---|
| Molecular Formula | C11H21BrO2 |
| Molecular Weight (g/mol) | 265.19 |
| MDL Number | MFCD00002732 |
| InChI Key | IUDGNRWYNOEIKF-UHFFFAOYSA-N |
| Synonym | undecanoic acid, 11-bromo, 11-bromodecanoic acid, 11-bromo-undecanoic acid, 11-bromoundecanoic acid,tech, 11-bromoundecanoicacid, 11bromoundecanoic acid, 11-bromohendecanoic acid, 11-bromo undecanoic acid, ksc489q1h, 11-bromoundecanoic acid |
| PubChem CID | 17812 |
| IUPAC Name | 11-bromoundecanoic acid |
| SMILES | C(CCCCCBr)CCCCC(=O)O |
Chemical Identifiers
| 2834-05-1 | |
| 265.19 | |
| IUDGNRWYNOEIKF-UHFFFAOYSA-N | |
| 17812 | |
| C(CCCCCBr)CCCCC(=O)O |
| C11H21BrO2 | |
| MFCD00002732 | |
| undecanoic acid, 11-bromo, 11-bromodecanoic acid, 11-bromo-undecanoic acid, 11-bromoundecanoic acid,tech, 11-bromoundecanoicacid, 11bromoundecanoic acid, 11-bromohendecanoic acid, 11-bromo undecanoic acid, ksc489q1h, 11-bromoundecanoic acid | |
| 11-bromoundecanoic acid |
Specifications
| 2834-05-1 | |
| White-Yellow | |
| C11H21BrO2 | |
| 25 g | |
| IUDGNRWYNOEIKF-UHFFFAOYSA-N | |
| 11-bromoundecanoic acid | |
| 17812 | |
| ≥97.0% (GC,T) | |
| 11-Bromoundecanoic Acid |
| 49°C | |
| 190°C | |
| MFCD00002732 | |
| undecanoic acid, 11-bromo, 11-bromodecanoic acid, 11-bromo-undecanoic acid, 11-bromoundecanoic acid,tech, 11-bromoundecanoicacid, 11bromoundecanoic acid, 11-bromohendecanoic acid, 11-bromo undecanoic acid, ksc489q1h, 11-bromoundecanoic acid | |
| C(CCCCCBr)CCCCC(=O)O | |
| 265.19 | |
| 265.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-2652
TSCA : Yes