missing translation for 'onlineSavingsMsg'
Learn More

11-Bromoundecanoic Acid, 98%

Catalog No. AC423160250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423160250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC423160250 Supplier Thermo Scientific Chemicals Supplier No. 423160250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2834-05-1 | C11H21BrO2 | 265.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2834-05-1
Molecular Formula C11H21BrO2
Molecular Weight (g/mol) 265.19
MDL Number MFCD00002732
InChI Key IUDGNRWYNOEIKF-UHFFFAOYSA-N
Synonym undecanoic acid, 11-bromo, 11-bromodecanoic acid, 11-bromo-undecanoic acid, 11-bromoundecanoic acid,tech, 11-bromoundecanoicacid, 11bromoundecanoic acid, 11-bromohendecanoic acid, 11-bromo undecanoic acid, ksc489q1h, 11-bromoundecanoic acid
PubChem CID 17812
IUPAC Name 11-bromoundecanoic acid
SMILES C(CCCCCBr)CCCCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48°C to 51°C
Color White to Beige
Boiling Point 173°C to 174°C (2.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)10CO2H
Quantity 25 g
Beilstein 02, II, 315
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols and hydrocarbons
Formula Weight 265.19
Percent Purity 98%
Physical Form Crystalline Chunks
Chemical Name or Material 11-Bromoundecanoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-602-9

RUO – Research Use Only