missing translation for 'onlineSavingsMsg'
Learn More

11-Bromo-1-undecanol 97.0+%, TCI America™

Catalog No. B14775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14775G 5 g
B147725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14775G Supplier TCI America Supplier No. B14775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1611-56-9
Molecular Formula C11H23BrO
Molecular Weight (g/mol) 251.21
MDL Number MFCD00004752
InChI Key XFGANBYCJWQYBI-UHFFFAOYSA-N
Synonym 11-bromo-1-undecanol, 11-bromoundecanol, 1-undecanol, 11-bromo, 1-bromo-11-hydroxyundecane, undecamethylene bromohydrin, 11-bromoundecyl alcohol, 11-bromo-l-undecanol, 11-bromoundecane-1-ol, 11-bromanylundecan-1-ol, acmc-1br3a
PubChem CID 74163
IUPAC Name 11-bromoundecan-1-ol
SMILES OCCCCCCCCCCCBr

Specifications

Melting Point 45°C
Boiling Point 170°C
Quantity 5 g
Formula Weight 251.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 11-Bromo-1-undecanol
TSCA No
Recommended Storage Refrigerator