Learn More
Fine Science Tools 11.5 CM BLUNT/BLUNTTISSUE
Catalog No. NC9749963
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 23mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No
Catalog No. NC9749963 Supplier Fine Science Tools Supplier No. 1407210
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 23mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No