missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11,12-Dihydro-11-phenylindolo[2,3-a]carbazole 97.0+%, TCI America™
$182.68 - $546.17
Chemical Identifiers
| CAS | 1024598-06-8 |
|---|---|
| Molecular Formula | C24H16N2 |
| Molecular Weight (g/mol) | 332.41 |
| MDL Number | MFCD22581305 |
| InChI Key | IENOQUQIVSMWGP-UHFFFAOYSA-N |
| Synonym | 11-Phenylindolo[2,3-a]carbazole |
| PubChem CID | 59365871 |
| IUPAC Name | 11-phenyl-11H,12H-indolo[2,3-a]carbazole |
| SMILES | N1C2=CC=CC=C2C2=C1C1=C(C=C2)C2=C(C=CC=C2)N1C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4860200MG
|
TCI America
D4860200MG |
200 mg |
Each for $182.68
|
|
|||||
|
D48601G
|
TCI America
D48601G |
1 g |
Each for $546.17
|
|
|||||
Chemical Identifiers
| 1024598-06-8 | |
| 332.41 | |
| IENOQUQIVSMWGP-UHFFFAOYSA-N | |
| 59365871 | |
| N1C2=CC=CC=C2C2=C1C1=C(C=C2)C2=C(C=CC=C2)N1C1=CC=CC=C1 |
| C24H16N2 | |
| MFCD22581305 | |
| 11-Phenylindolo[2,3-a]carbazole | |
| 11-phenyl-11H,12H-indolo[2,3-a]carbazole |
Specifications
| 1024598-06-8 | |
| C24H16N2 | |
| 200 mg | |
| IENOQUQIVSMWGP-UHFFFAOYSA-N | |
| 11-phenyl-11H,12H-indolo[2,3-a]carbazole | |
| 59365871 | |
| ≥97.0% (HPLC,N) | |
| 11,12-Dihydro-11-phenylindolo[2,3-a]carbazole |
| White-Yellow | |
| MFCD22581305 | |
| 11-Phenylindolo[2,3-a]carbazole | |
| N1C2=CC=CC=C2C2=C1C1=C(C=C2)C2=C(C=CC=C2)N1C1=CC=CC=C1 | |
| 332.41 | |
| 332.41 | |
| Crystalline Powder |
Safety and Handling
TSCA : No