missing translation for 'onlineSavingsMsg'
Learn More

Allantoin, USP, 98.5-101%, Spectrum™ Chemical

Catalog No. 18600403
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600403 12 kg Fiber Drum
18600400 100 g Fiber Drum
18600401 500 g Fiber Drum
18600402 2.5 kg Fiber Drum
18600399 25 kg Fiber Drum
18600398 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18600403 Supplier Spectrum Chemical Mfg Cor Supplier No. A272112KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A2721, 97-59-6, C4H6N4O3

Spectrum™ Chemical Allantoin, USP promotes cell proliferation and wound healing and has a soothing, anti-irritant, and skin protectant effect. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 97-59-6
Molecular Formula C4H6N4O3
Molecular Weight (g/mol) 158.12
InChI Key POJWUDADGALRAB-UHFFFAOYNA-N
IUPAC Name (2,5-dioxoimidazolidin-4-yl)urea
SMILES NC(=O)NC1NC(=O)NC1=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.1%
Packaging Fiber Drum
Quantity 12 kg
Percent Purity 98.5 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Allantoin