missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cupric Hydroxide, Technical, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Powder, Needles or Crystals |
| Quantity | 500 g |
| Molecular Weight (g/mol) | 97.56 |
| Grade | Technical |
| Packaging | Amber Glass Bottle |
| Chemical Name or Material | Cupric Hydroxide |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.