Learn More
Celltreat Scientific Products 10mL Wobble-not Serological Pipet, Individually Wrapped, Paper/Plastic, Bag, Sterile
Catalog No. NC2206329
Encompass
No Wobble. No Drip. Wobble-not serological pipets require less insertion force due to the patented, innovative 2-tier step design. Wobble-nots are the only serological pipets that will give you two zones of contact in your pipet controllers nozzle, to increase stability and reduce dripping. Ultra-clear markings and color-coded stripes for quick volume identification. Accuracy 2% at full volume. Ideal liquid handling partner when paired with ali-Q™ 2 Pipet Controller.
Catalog No. NC2206329 Supplier Celltreat Scientific Products Supplier No. 229092B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
No Wobble. No Drip. Wobble-not serological pipets require less insertion force due to the patented, innovative 2-tier step design. Wobble-nots are the only serological pipets that will give you two zones of contact in your pipet controllers nozzle, to increase stability and reduce dripping. Ultra-clear markings and color-coded stripes for quick volume identification. Accuracy 2% at full volume. Ideal liquid handling partner when paired with ali-Q™ 2 Pipet Controller.