missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LaMotte™ 10mL Square Test Tube
For use with LaMotte™ water quality monitoring kits.
Supplier: LaMotte™ 0106EMD
Specifications
| Test Tube | |
| Plastic | |
| Used in LaMotte™ water quality monitoring kits |
| 10 mL | |
| Each |