CELLTREAT Scientific Products 10mL Open End Serological Pipet. Wide, open end is ideal for viscous and high particulate solutions, and slurries such as Sephadex. All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in paper/plastic wrappers. 50/bag, 200/case.