Learn More
Celltreat Scientific Products 10mL Serological Pipet, Paper/Plastic Wrapped, Sterile
Catalog No. NC1772269
Encompass
CELLTREAT Scientific Products 10mL Serological Pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Now available in double case quantities. Double case serological pipets offer a price savings. Save up to 65% on shipping costs. Reduce your order frequency and cardboard waste.
Catalog No. NC1772269 Supplier Celltreat Scientific Products Supplier No. 229011B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CELLTREAT Scientific Products 10mL Serological Pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934. Bi-directional (positive and negative) graduations are printed in bold easy-to-read ink. Premium material barrier plug. Color coded stripes and packaging enable quick volume identification. Printed indicator arrow for quickly locating negative graduations when using a pipet controller. Now available in double case quantities. Double case serological pipets offer a price savings. Save up to 65% on shipping costs. Reduce your order frequency and cardboard waste.