missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 10 mL Nalgene Oak Ridge Tube or 12mL Round Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor

Catalog No. 0101311
Catalog No. 0101311 Supplier Thermo Scientific™ Supplier No. 0101311

Accommodate 10mL Oak Ridge or 12mL round tubes in Thermo Scientific™ Fiberlite™ conical tube rotors.

Accommodate 10mL Nalgene Oak Ridge or 12mL round tubes in Thermo Scientific™ Fiberlite™ F13-14x50cy or Fiberlite F15-8x50cy rotors.

Specifications

Type Ridge Tube Adapter
Tube Type 10 mL Nalgene Oak Ridge Tube, 12 mL Round Tube
Capacity 10, 12 mL
For Use With (Equipment) Fiberlite F13-14x50cy, Fiberlite F15-8x50cy rotors
Description 10mL Nalgene Oak Ridge Tube or 12mL Round Tube Adapter