missing translation for 'onlineSavingsMsg'
Learn More

Lactose, Monohydrate, BiotechGrade, Spectrum™ Chemical

Catalog No. 18603693
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603693 45 kg Poly Bottle
18603690 500 g Poly Bottle
18603691 2.5 kg Poly Bottle
18603692 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603693 Supplier Spectrum Chemical Mfg Cor Supplier No. L103745KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L1037, 64044-51-5, C12H22O11·H2O

Spectrum™ Chemical Lactose, Monohydrate, BiotechGrade , also known as milk sugar, is a disaccharide sugar and is added to pills as a filler because of its physical properties. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 64044-51-5
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
InChI Key WSVLPVUVIUVCRA-RJMJUYIDSA-N
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol hydrate
SMILES O.OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Physical Form Crystalline Powder or Needles
Quantity 45 kg
Grade Biotech
Loss on Drying 0.5%
Packaging Poly Bottle
Appearance Fine, white powder
Chemical Name or Material Lactose, Monohydrate