Learn More
Chemglass Inc 10000mL Reaction Vessel, Cylindrical, 200mm Schott O-Ring Flange, 215mm O.D. x 440mm Height
Catalog No. CG192006
Encompass
10000mL Reaction Vessel, Cylindrical, 200mm Schott O-Ring Flange, 215mm O.D. x 440mm Height. Cylindrical reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. For clamp see CG-141 or CG-141-T, for o-ring see CG-147.
Catalog No. CG192006 Supplier Chemglass Inc Supplier No. CG192006
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10000mL Reaction Vessel, Cylindrical, 200mm Schott O-Ring Flange, 215mm O.D. x 440mm Height. Cylindrical reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. For clamp see CG-141 or CG-141-T, for o-ring see CG-147.