Learn More
Foxx Life Sciences EZBio Single Use Carboy (Bottle) Assembly, 10L (2.5GAL) Gamma Sterilized Light Blocking Amber HDPE Carboy for Aseptic Transfer and Sterile Storage with VersaCap 83B (83mm), Vent and Dip Tube, 1/EA
GREENER_CHOICE
Catalog No. NC1796834
Encompass
EZBio Gamma Sterilized Carboys are with distinctive rectangular shape saves valuable bench space while the molded grips provide a secure hold to facilitate in lifting, handling, and pouring. With easy to read metric graduation marks, certified to 5% accuracy, these carboys include pre-mounted, high tensile strength, platinum-cured silicone tubing. This assembly provides a flexible two-port filling and venting system, allowing for extractable free, aseptic gas and liquid transfer as required for pharmaceutical, biotechnology, and laboratory fluid transfer applications. These high-quality carboy assemblies are readily able to accept additional fluid transfer components such as aseptic connectors, filters, and manifold integration. Gamma Irradiated in the United States.USP Class VI, FDA Grade materialsHigh-impact strengthErgonomic shape Leakproof in ID TPE Tubing
Catalog No. NC1796834 Supplier Foxx Life Sciences Supplier No. 55KGB01FLS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
EZBio Gamma Sterilized Carboys are with distinctive rectangular shape saves valuable bench space while the molded grips provide a secure hold to facilitate in lifting, handling, and pouring. With easy to read metric graduation marks, certified to 5% accuracy, these carboys include pre-mounted, high tensile strength, platinum-cured silicone tubing. This assembly provides a flexible two-port filling and venting system, allowing for extractable free, aseptic gas and liquid transfer as required for pharmaceutical, biotechnology, and laboratory fluid transfer applications. These high-quality carboy assemblies are readily able to accept additional fluid transfer components such as aseptic connectors, filters, and manifold integration. Gamma Irradiated in the United States.USP Class VI, FDA Grade materialsHigh-impact strengthErgonomic shape Leakproof in ID TPE Tubing