Learn More
Beckman Coulter Biomek Span-8 P1000 Conductive Tips, with Barrier
Catalog No. NC9491796
Encompass
Conductive tips for use with Span-8 Pod to enable Liquid Level Sensing
Catalog No. NC9491796 Supplier Beckman Coulter Supplier No. BEC987925
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Conductive tips for use with Span-8 Pod to enable Liquid Level Sensing