Learn More
Celltreat Scientific Prod 100mm x 15mm Petri Dish, 2 Compartments, Sterile DFS Item
Supplier:  Celltreat Scientific Products 229682
CELLTREAT Scientific Products 2 Compartment Petri Dish. Designed for simultaneous studies of organisms in a common environment. Molded dividers create completely separate compartments while allowing airflow. Ridge on bottom dish allows easy handling and stacking. Lids contain molded spacers to allow gas exchange. Two compartments with working volume of 7-8mL and growth area of 28.3cm. Lid contains stacking rings for increased stability. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/bag, 500/case.
Catalog No. 50828745
Only null left
Explore available promotions