Learn More
Celltreat Scientific Products 100mm x 15mm Petri Dish, Stackable, Sterile
Supplier:  Celltreat Scientific Products 229695
Encompass
CELLTREAT Scientific Products 100mm x 15mm Stackable Petri Dish. Non-Treated Petri Dish. CELLTREAT Petri Dishes are manufactured in high capacity automated production lines resulting in high quality, value priced products. Ventilation ribs reduce condensation and allow air circulation. Consistent flatness allows for even media distribution. Lids contain stacking rings for increased stability. Working volume of 15-16mL and growth area of 55cm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 20/bag, 500/case.
Catalog No. 50828750
Only null left