No Wobble. No Drip. Wobble-not serological pipets require less insertion force due to the patented, innovative 2-tier step design. Wobble-nots are the only serological pipets that will give you two zones of contact in your pipet controllers nozzle, to increase stability and reduce dripping. Ultra-clear markings and color-coded stripes for quick volume identification. Accuracy 2% at full volume. Ideal liquid handling partner when paired with ali-Q™ 2 Pipet Controller.