Learn More
Chemglass Inc 100mL Glas-Col Heating Mantle, Series TM, Spherical, 80 Watts
Catalog No. CG1200002
Encompass
100mL Glas-Col Heating Mantle, Series TM, Spherical, 80 Watts. The Glas-Col Series TM spherical flask mantle covers only the flasks bottom half, letting you see its full contents. The Series TM mantle is available with a bottom hole for flask drain on a special order basis. Terminal box models are available with switches and pilot lights. Mantles are 115 Volt.
Catalog No. CG1200002 Supplier Chemglass Inc Supplier No. CG1200002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
100mL Glas-Col Heating Mantle, Series TM, Spherical, 80 Watts. The Glas-Col Series TM spherical flask mantle covers only the flasks bottom half, letting you see its full contents. The Series TM mantle is available with a bottom hole for flask drain on a special order basis. Terminal box models are available with switches and pilot lights. Mantles are 115 Volt.