100mL Heavy Wall Pressure Vessel, Cylindrical, #15 Internal Thread, Vessel Only, 38mm O.D. x 178mm Length, Bushing Size/15. Heavy wall pressure vessels have an internal thread for use with PTFE bushings as pressure seals. The design allows hand tightening/loosening for easy access into the vessel. O-ring is positioned on the bushing above the thread for improved sealing. Supplied complete with vessel, PTFE bushing & Viton o-ring. Optional Perfluoro o-ring has the chemical resistance & service temperature range similar to Kalrez 4079. See CG-362, CG-364, and CG-366 for optional PTFE Bushings.IMPORTANT NOTICE: ALL CHEMGLASS PRESSURE VESSELS ARE TESTED AT 150 PSI, OR APPROX TWO AND ONE-HALF TIMES THEIR RECOMMENDED WORKING PRESSURE. HOWEVER, CHEMGLASS DOES NOT GUARANTEE THE VESSEL FROM BREAKING UNDER PRESSURE, DUE TO THE NATURE OF MATERIAL AND CONDITIONS BEYOND OUR CONTROL. CHEMGLASS RECOMMENDS THE USE OF SAFETY SHIELDS AND/OR OTHER SAFETY EQUIPMENT.