Learn More
Haws™ Foots Treadle Assembly
Catalog No. 17985555
Change view
Click to view available options
For Use With (Equipment):
Pedestal and Combination Unit
Catalog No. For Use With (Equipment)
17985555 Pedestal and Combination Unit
1 options
Catalog No. 17985555 Supplier Haws™ Supplier No. SP 220
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Model SP220 foot control assembly, including powder-coated aluminum treadle for hands free activation of eyewash.
Model SP220 is a complete foot control assembly, including chain, clamp, nut, ring, screw, spring and treadle. For use on pedestal and combination units.
Specifications
For Use With (Equipment) Pedestal and Combination Unit
Includes Chain, Clamp, Nut, Ring, Screw, Spring and Treadle
Product Type Foot Treadle Assembly