Learn More
Brady™ i7500 Printer Accessory Label Taken Sensor, Black
Description
- When printing in peel mode, the sensor automatically triggers the next label print when the currently-presented label is removed.
- Location of the detection sensor can be adjusted cross-web across the material print path.
- Peel results vary depending on auto-dispense capability of material and label size; user should test on intended material and application.
Specifications
Specifications
| Product Type | Sensor |
| For Use With (Equipment) | i7500 |