Learn More
Inorganic Ventures 1000g/mL ANTIMONY 125mL
Catalog No. 50723406
Encompass
Inorganic Ventures' 1000 ppm (g/mL) Antimony for ICP is a 125mL certified reference material set in a nitric acid / tartaric acid matrix for stability. This antimony standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50723406 Supplier Inorganic Ventures Supplier No. CGSB1125ML
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Inorganic Ventures' 1000 ppm (g/mL) Antimony for ICP is a 125mL certified reference material set in a nitric acid / tartaric acid matrix for stability. This antimony standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search