Learn More
Chemglass Inc 1000mL Reaction Vessel, Jacketed, 100mm Schott O-Ring Flange, 3/4"" Beaded Pipe Inlet/Outlet, CG-911-A-10 Drain Valve
Catalog No. 50872352
Encompass
1000mL Reaction Vessel, Jacketed, 100mm Schott O-Ring Flange, 3/4" Beaded Pipe Inlet/Outlet, CG-911-A-10 Drain Valve. Features: Chemically Resistant Perfluoro O-Rings On Drain Valves Graduation Scale On Both Sides Showing Approximate Volume Sidearm on Lower Drain Valve Can Be Used With a Standard 1/2 or 3/4in TFE, PFA or PP Compression Fitting All Vessels Are Serial Numbered GL Thread Inlet/Outlets Are Supplied With Removable High Temperature PFA Hose Connections And PPS Screw Caps 3/4in Beaded Pipe Inlet/Outlets Are For Use With CG-1968-70 Couplings, CG-1969-A-03 Or CG-1964-42 Adapters
Catalog No. 50872352 Supplier Chemglass Inc Supplier No. CG192915
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
1000mL Reaction Vessel, Jacketed, 100mm Schott O-Ring Flange, 3/4" Beaded Pipe Inlet/Outlet, CG-911-A-10 Drain Valve. Features: Chemically Resistant Perfluoro O-Rings On Drain Valves Graduation Scale On Both Sides Showing Approximate Volume Sidearm on Lower Drain Valve Can Be Used With a Standard 1/2 or 3/4in TFE, PFA or PP Compression Fitting All Vessels Are Serial Numbered GL Thread Inlet/Outlets Are Supplied With Removable High Temperature PFA Hose Connections And PPS Screw Caps 3/4in Beaded Pipe Inlet/Outlets Are For Use With CG-1968-70 Couplings, CG-1969-A-03 Or CG-1964-42 Adapters