Learn More
Chemglass Inc 1000mL Glassblowers RBF Blank
Catalog No. CG61810
Encompass
1000mL Glassblowers RBF Blank. Blanks for fabrication of single or multi-neck round bottom flasks. NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. CG61810 Supplier Chemglass Inc Supplier No. CG61810
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
1000mL Glassblowers RBF Blank. Blanks for fabrication of single or multi-neck round bottom flasks. NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.